Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphotungstic acid hydrate

Catalog No. 45597050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-597-050G 50 g
45-597-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-597-050G Supplier Sigma Aldrich Supplier No. 45597050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphotungstic acid hydrate

Specifications

Melting Point ∼95°C
CAS 12501-23-4
Quantity 50 g
Molecular Formula H3O40PW12 -+ xH2O
Linear Formula H3[P(W3O10)4 ] -+ xH2O
MDL Number MFCD00149915
Synonym Tungstophosphoric acid
Molecular Weight (g/mol) 2880.05
Percent Purity 99.995% trace metals basis (Purity excludes up to 300 ppm Si)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.