Learn More
Abcam Phosphotungstic acid hydrate, Histology stain for cell specimens, 25G
Catalog No. 502704844
Encompass_Preferred
MW 2880.17 g/mol. Histology stain for cell specimens. Heteropoly acid that may be used as a staining agent for transmission electron microscope (TEM) imaging.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4844 Supplier Abcam Supplier No. AB14620625G
MW 2880.17 g/mol. Histology stain for cell specimens. Heteropoly acid that may be used as a staining agent for transmission electron microscope (TEM) imaging.
The product is subject to the following: Abcam Restricted Use Statement