Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotungstic Acid TS, Ricca Chemical
SDP

Catalog No. 5859616 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58596-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58596-16 Supplier Ricca Chemical Company Supplier No. 5859.616
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 12501-23-4
Molecular Weight (g/mol) 2880.2 g/mol

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Phosphotungstic Acid TS
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.