Learn More
Enzo Life Sciences Phosphotyrosine monoclonal antibody (PY20) (agarose immobilized) (0.5ML)
SDP
Supplier:  Enzo Life Sciences BMLSA2410500
Encompass_Preferred
Covalently coupled to 4% cross-linked agarose beads. This product is optimized for binding of phosphotyrosine containing proteins with minimal non-specific binding. Clone: PY20. Isotype: IgG2b. Immunogen: Phosphotyrosine. Source: Purified from mouse ascites. Species reactivity: Species independent. Applications: IP Immunoaffinity chromatography. Formulation: 50% slurry (0.5ml bed volume, 1ml total volume) at a conjugation ratio of 8-10 mg of PY20 per ml of packed beads.
Catalog No. 50-201-1775