Learn More
Enzo Life Sciences Phosphotyrosine monoclonal antibody (PY20) (biotin conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2430100
Encompass_Preferred
Conjugated to biotin. Clone: PY20. Isotype: IgG2b. Immunogen: Phosphotyrosine. Species reactivity: Species independent. Applications: ELISA, ICC, IHC. Purity: Affinity Purified. Formulation: Liquid. In 20mM sodium phosphate containing 150mM NaCl, 50% glycerol, and 3mM sodium azide, pH 7.5.
Catalog No. 50-201-1776