Learn More
ECM Bioscience Anti-Phosphotyrosine Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP2221
Encompass_Preferred
This antibody detects phosphotyrosine-containing proteins on SDS-PAGE immunoblots of EGF treated A431 cells, as well as pervanadate treated A431, HeLa, Jurkat, and endothelial cells. The immunogen was Anti-Phosphotyrosine was generated from a panel of phosphotyrosine-containing peptide immunogens designed from human protein sequences. All peptide sequences used are highly conserved phosphotyrosine motifs. Accession #. Applications = WB, E, IP, ICC; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = phosphotyrosine, phoshoserine, phoshothreonine
Catalog No. 50-172-0854