Learn More
Sigma Aldrich Fine Chemicals Biosciences PhosSTOP, 10T
Catalog No. NC0962508
Encompass
Synonym: inhibitor tablets for phosphatase, phosphatase inhibitor tablets. PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).
Catalog No. NC0962508 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4906845001
Synonym: inhibitor tablets for phosphatase, phosphatase inhibitor tablets. PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).