Learn More
Medchemexpress LLC PhosTAC7 | 3016307-75-5 | 99.23% | C58H87ClN2O17 | 50 MG
SDP
Catalog No. 5000353272
Encompass_Preferred
PhosTAC7 is a heterobifunctional molecule, also known as a Phosphorylation Targeting Chimera (PhosTAC). It is designed to dephosphorylate specific proteins by recruiting serine/threonine protein phosphatase 2A (PP2A), making it a valuable tool for various research applications.
  • Selectively modulates the phosphorylation state of individual target proteins.
  • Dephosphorylates PDCD4, FOXO3a, and tau proteins.
  • Useful in research for cancer.
  • Applicable in the study of tau protein-related neurodegenerative diseases, such as Alzheimer's.
  • For research use only.
Catalog No. 50-003-53272 Supplier Medchemexpress LLC Supplier No. HY14523250MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PhosTAC7 is a heterobifunctional molecule, also known as a Phosphorylation Targeting Chimera (PhosTAC). It is designed to dephosphorylate specific proteins by recruiting serine/threonine protein phosphatase 2A (PP2A), making it a valuable tool for various research applications.
  • Selectively modulates the phosphorylation state of individual target proteins.
  • Dephosphorylates PDCD4, FOXO3a, and tau proteins.
  • Useful in research for cancer.
  • Applicable in the study of tau protein-related neurodegenerative diseases, such as Alzheimer's.
  • For research use only.