Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM2793P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: PY793. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: MCF-7, MDA-231, T47-D cells or breast carcinoma. Cellular Staining: Depends upon the location of phosphorylated target. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Phosphotyrosine conjugated to BSA. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5457