Learn More
Hach Company PhosVer 3 Phosphate Reagent AccuVac Ampules, pk/25
Supplier:  Hach Company 2508025
Encompass
For determination of reactive Phosphorus (Phosphate) by the ascorbic acid method, using PhosVer 3 Phosphate Reagent AccuVac Ampules. USEPA accepted for reporting for wastewater analysis using Hach Method 8048. Range: 0.02 - 2.50 mg/L as PO4 Pack of 25 ampules.
Catalog No. 50-199-6115