Learn More
Hach Company PhosVer 3 Phosphate Reagent Powder Pillows, 10 mL, pk/50
Catalog No. 501995403 Shop All Hach Company Products
Encompass
For determination of reactive (ortho) phosphate (phosphorus) by the PhosVer 3 ascorbic acid method. USEPA accepted for reporting for wastewater analysis using Hach Method 8048, Phosphorus, Reactive. Sample Size: 10 mL. Also used in acid-hydrolyzable and total phosphorus determination, after appropriate digestion. Pack of 50 powder pillows.
Catalog No. 50-199-5403 Supplier Hach Company Supplier No. 2106046
For determination of reactive (ortho) phosphate (phosphorus) by the PhosVer 3 ascorbic acid method. USEPA accepted for reporting for wastewater analysis using Hach Method 8048, Phosphorus, Reactive. Sample Size: 10 mL. Also used in acid-hydrolyzable and total phosphorus determination, after appropriate digestion. Pack of 50 powder pillows.