Learn More
Hach Company PhosVer 3 Phosphate Reagent Powder Pillows, 5 mL, pk/100
Catalog No. 501994722 Shop All Hach Company Products
Encompass
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method. Sample Size: 5 mL. For use with visual test kits. Pack of 100 powder pillows.
Catalog No. 50-199-4722 Supplier Hach Company Supplier No. 220999
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method. Sample Size: 5 mL. For use with visual test kits. Pack of 100 powder pillows.