Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosvitin from egg yolk50MG
Catalog No. 502901997
Encompass
Phosvitin from egg yolk is a polyanionicnbspphosphoserine rich protein. This phosphoglycoprotein comprises helices and -sheets structural elements which are poorly organized due to the presence of phosphoserine. It represents about 11% of yolk proteins and is hydrophilic.
Catalog No. 50-290-1997 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P125350MG
Phosvitin from egg yolk is a polyanionicnbspphosphoserine rich protein. This phosphoglycoprotein comprises helices and -sheets structural elements which are poorly organized due to the presence of phosphoserine. It represents about 11% of yolk proteins and is hydrophilic.