Learn More
Electron Microscopy Sciences Photo Etched Coverslips 18x18 mm
SDP
Catalog No. 50949526
Encompass
These square gridded coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. Using No. 2 (.17 -.24 mm) thickness cover slips. Grid contains 520 alphanumeric coded squares, each square 600 x 600 um (0.6 mm x0.6 mm). Used to facilitate mounting of micromaniputated cells or chromosome spreads.
Catalog No. 50-949-526 Supplier Electron Microscopy Sciences Supplier No. 7226418
These square gridded coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. Using No. 2 (.17 -.24 mm) thickness cover slips. Grid contains 520 alphanumeric coded squares, each square 600 x 600 um (0.6 mm x0.6 mm). Used to facilitate mounting of micromaniputated cells or chromosome spreads.