Learn More
Electron Microscopy Sciences Photo Etched Coverslips 12 mm
SDP
Catalog No. 50949642
Encompass
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. 25 mm diameter. Using #2 (0.17 0.25 mm) thickness cover slips. Unique grid pattern, numbered 1 to 200 squares, each 500 um width. Suitable for insertion into most 6-well plates. Adaptable to most microscope mounting plates. Facilitates observation of activity growing cells.
Catalog No. 50-949-642 Supplier Electron Microscopy Sciences Supplier No. 7226512
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. 25 mm diameter. Using #2 (0.17 0.25 mm) thickness cover slips. Unique grid pattern, numbered 1 to 200 squares, each 500 um width. Suitable for insertion into most 6-well plates. Adaptable to most microscope mounting plates. Facilitates observation of activity growing cells.