Learn More
Electron Microscopy Sciences Photo Etched German Glass Coverslips, 25 mm
SDP
Catalog No. 50949634
Encompass
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. 25 mm diameter. Using #2 (0.17– 0.25 mm) thickness cover slips. Unique grid pattern, numbered 1 to 200 squares, each 500 um width. Suitable for insertion into most 6-well plates. Adaptable to most microscope mounting plates. Facilitates observation of activity growing cells.
Catalog No. 50-949-634 Supplier Electron Microscopy Sciences Supplier No. 7226550
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern. 25 mm diameter. Using #2 (0.17– 0.25 mm) thickness cover slips. Unique grid pattern, numbered 1 to 200 squares, each 500 um width. Suitable for insertion into most 6-well plates. Adaptable to most microscope mounting plates. Facilitates observation of activity growing cells.