Learn More
Motic™ Photo Eyepieces
SCI_ED
Catalog No. 01674243 Shop All Motic Group Products
Change view
Click to view available options
Magnification Power:
2.5X
4X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
01-674-243 4X
01-674-242 2.5X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-674-243 Supplier Motic™ Supplier No. 1101000300452
Add to Cart
Add to Cart
Photo eyepiece to be used with the C-mount Photo Eyepiece Adapter.
  • Also requires C-mount Photo Eyepiece Adapter and T-Mount adapter from your camera's manufacturer
Specifications
For Use With (Application) Attaching DSLR Camera To Microscope
For Use With (Equipment) BA210S/E Series, BA310E/POL/MET Series, BA410E Series, Panthera C2/E2/Cloud/TEC MET/TEC POL Series, PA43 BIO Series, AE31E/2000/2000 MET Series, SMZ171 Series and GM171 Series
Magnification Power 4X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov