Learn More
ACCU-SCOPE™ Photo Tube
SDP
Catalog No. 01189298 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
Video Adapter
Catalog No. Type
01-189-298 Video Adapter
1 options
Catalog No. 01-189-298 Supplier ACCU-SCOPE™ Supplier No. 002010P
Only null left
Add to Cart
Add to Cart
ACCU-SCOPE Microscope photo tube, 23 mm I.D, with focus adjustment. Requires c-mount camera adapter (CAT # 2005-5M).
  • Photo tube with focus adjustment
  • Requires c-mount camera adapter (CAT # 2005-5M)
Specifications
Type Video Adapter
Autoclavable Not Autoclavable
Sterility Non-sterile