Learn More
Advanced Biomatrix PhotoAlginate Methacrylated Alginate 200 mg sterile
Catalog No. 502255921
Encompass_Preferred
PhotoAlginate is a sterile, lyophilized, methacrylated alginate. It creates 3D hydrogels that can be prepared at multiple concentrations and crosslinked using light or calcium to provide various gel stiffness. PhotoAlginate can be mixed with other photocrosslinkable ECM's.
This item is not returnable. View return policy
Catalog No. 50-225-5921 Supplier Advanced Biomatrix Supplier No. 5310200MG
PhotoAlginate is a sterile, lyophilized, methacrylated alginate. It creates 3D hydrogels that can be prepared at multiple concentrations and crosslinked using light or calcium to provide various gel stiffness. PhotoAlginate can be mixed with other photocrosslinkable ECM's.