Learn More
Advanced Biomatrix PhotoAlginate Methacrylated Alginate 200 mg sterile with LAP kit
Supplier:  Advanced Biomatrix 53541KIT
Encompass_Preferred
PhotoAlginate is a sterile, lyophilized, methacrylated alginate. It creates 3D hydrogels that can be prepared at multiple concentrations and crosslinked using light or calcium to provide various gel stiffness. PhotoAlginate can be mixed with other photocrosslinkable ECM's.
Catalog No. 50-225-5946