Learn More
Ace Glass, Inc. Photochem reactor body, 1000mL, complete with spinbar, sparger, bushings and PTFE tubing
SDP
Catalog No. 50975801
Encompass_Preferred
1000mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.
Catalog No. 50-975-801 Supplier Ace Glass, Inc. Supplier No. 786340
1000mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.