Learn More
Ace Glass, Inc. Photochem reactor body, 500mL, complete with spinbar, sparger, bushings and PTFE tubing
SDP
Catalog No. 50979588
Encompass_Preferred
500mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.
Catalog No. 50-979-588 Supplier Ace Glass, Inc. Supplier No. 786338
500mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.