Learn More
Ace Glass, Inc. Photochem reactor body, 500mL, complete with spinbar, sparger, bushing and PTFE tubing
SDP
Catalog No. 50975514
Encompass_Preferred
Complete photochemical reactor featuring spinbar, sparger, bushing and PTFE tubing. 500mL reaction vessel is made of borosilicate glass to accomm ODate ACE 7854, 7856, or 7857 immersion wells. Vessel has one 14/20 angled joint for sparger tube, one 24/40 vertical joint for condenser, and one #7 Ace-Thred side arm for a thermometer. Center joint is 60/40, and the bottom of the reactor is flat to allow use of a stir magnet.
Catalog No. 50-975-514 Supplier Ace Glass, Inc. Supplier No. 784115
Complete photochemical reactor featuring spinbar, sparger, bushing and PTFE tubing. 500mL reaction vessel is made of borosilicate glass to accomm ODate ACE 7854, 7856, or 7857 immersion wells. Vessel has one 14/20 angled joint for sparger tube, one 24/40 vertical joint for condenser, and one #7 Ace-Thred side arm for a thermometer. Center joint is 60/40, and the bottom of the reactor is flat to allow use of a stir magnet.