Learn More
Ace Glass, Inc. Photochemical reactor turntable without glass or filters, 120vac 50/60Hz DC motor control, 1/29hP DC motor, CE rated
SDP
Catalog No. 50975817
Encompass
Ace photochemical reactor turntable for determining relative and absolute quantum yields. Complete unit includes 1/29hP DC motor and 120vac 50/60Hz DC motor control. Variable height disc holds 13mm OD sample tubes: 33 in outer circle, 18 in inner circle. Unit will hold up to 4 ACE 7891 series square filters. CE rated. *UV lamp, power supply, immersion well, fiters, and sample tubes not included.
Catalog No. 50-975-817 Supplier Ace Glass, Inc. Supplier No. 789130
Add to Cart
Add to Cart
Request a Quote
Ace photochemical reactor turntable for determining relative and absolute quantum yields. Complete unit includes 1/29hP DC motor and 120vac 50/60Hz DC motor control. Variable height disc holds 13mm OD sample tubes: 33 in outer circle, 18 in inner circle. Unit will hold up to 4 ACE 7891 series square filters. CE rated. *UV lamp, power supply, immersion well, fiters, and sample tubes not included.