Learn More
Cayman Chemical PhotoclIck MyrIstIc AcId 1mg
SDP
Catalog No. 502932863
Encompass_Preferred
A derivative of myristic acid; contains a terminal clickable alkyne and a photoactivatable diazirine at carbon 10
Catalog No. 50-293-2863 Supplier Cayman Chemical Supplier No. 253631 MG
A derivative of myristic acid; contains a terminal clickable alkyne and a photoactivatable diazirine at carbon 10