Learn More
Cayman Chemical PhotoclIck MyrIstIc AcId 500ug
SDP
Catalog No. 502932711
Encompass_Preferred
A derivative of myristic acid; contains a terminal clickable alkyne and a photoactivatable diazirine at carbon 10
Catalog No. 50-293-2711 Supplier Cayman Chemical Supplier No. 25363500 UG
A derivative of myristic acid; contains a terminal clickable alkyne and a photoactivatable diazirine at carbon 10