Learn More
Cayman Chemical Photoclick Palmitic Acid, Purity: ≥95%, CAS: 2988174-40-7, 10 mg
SDP
Catalog No. 502919483
Encompass_Preferred
A derivative of palmitic acid; contains a terminal clickable alkyne moiety and a photoactivatable diazirine group at carbon 12
Catalog No. 50-291-9483 Supplier Cayman Chemical Supplier No. 3796910 MG
A derivative of palmitic acid; contains a terminal clickable alkyne moiety and a photoactivatable diazirine group at carbon 12