Learn More
Advanced Biomatrix PhotoDextran Methacrylated Dextran sterile 1 gram
Catalog No. 502255922
Encompass_Preferred
PhotoDextran is 1 gram of lyophilized methacrylated dextran. PhotoDextran provides 3D hydrogels that can be prepared at multiple concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. Can be mixed with other photocrosslinkable ECM's.
This item is not returnable. View return policy
Catalog No. 50-225-5922 Supplier Advanced Biomatrix Supplier No. 53111GM
PhotoDextran is 1 gram of lyophilized methacrylated dextran. PhotoDextran provides 3D hydrogels that can be prepared at multiple concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. Can be mixed with other photocrosslinkable ECM's.