Learn More
Bellco Glass, Inc. Photoetch Coverslip,12mm German Glass
SDP
Catalog No. 501944690
Encompass_Preferred
Ideal for florescence and con-focal microscopy Size: 12mm round od Material: German glass ( d-263m) Grid pattern: diamond pattern 100 alphanumeric coded squares Each square measures 0.6mm x 0.6mm
Catalog No. 50-194-4690 Supplier Bellco Glass, Inc. Supplier No. 191691012
Ideal for florescence and con-focal microscopy Size: 12mm round od Material: German glass ( d-263m) Grid pattern: diamond pattern 100 alphanumeric coded squares Each square measures 0.6mm x 0.6mm