Learn More
Bellco Glass, Inc. Photoetch Coverslip,18x18mm
SDP
Catalog No. 501944692
Encompass_Preferred
Ideal for florescence and confocal microscopy 520 alphanumeric coded squares .17-.19mm thick Each square measures 0.6mm x 0.6mm #1.5 thickness Facilitates mounting of micro-manipulated cells or chromosome spreads
Catalog No. 50-194-4692 Supplier Bellco Glass, Inc. Supplier No. 191691818
Ideal for florescence and confocal microscopy 520 alphanumeric coded squares .17-.19mm thick Each square measures 0.6mm x 0.6mm #1.5 thickness Facilitates mounting of micro-manipulated cells or chromosome spreads