Learn More
Bellco Glass, Inc. Photoetch Coverslip, 25mm German Glass
SDP
Catalog No. 501944691
Encompass_Preferred
Ideal for florescence and con-focal microscopy Size: 25mm round od Material: German glass ( d-263m) 200 areas in diamond pattern Each square measures 0.6mm x 0.6mm Facilitates mounting of micro-manipulated cells or chromosome spreads Line thickness: .02mm
Catalog No. 50-194-4691 Supplier Bellco Glass, Inc. Supplier No. 191691025
Ideal for florescence and con-focal microscopy Size: 25mm round od Material: German glass ( d-263m) 200 areas in diamond pattern Each square measures 0.6mm x 0.6mm Facilitates mounting of micro-manipulated cells or chromosome spreads Line thickness: .02mm