Learn More
Vernier™ Photogate Bar Tape Kit
SDPSCI_ED
Catalog No. S16262ND Shop All Vernier Software Products
Change view
Click to view available options
Length (Metric):
3 m
Catalog No. Length (Metric)
S16262ND 3 m
1 options
Catalog No. S16262ND Supplier Vernier™ Supplier No. TAPEVPG
Add to Cart
Add to Cart
Includes
Photogate Bar Tape Kit
The Photogate Bar Tape Kit contains a flexible plastic tape called Bar Tape and a guide that allows the tape to be used with a Vernier Photogate.
The tape is 3 m long, and it has alternating opaque and transparent bars that have a spacing of 1.524 cm. The tape can be attached to objects and pulled through the photogate. This allows you to perform experiments much like what you would do with a ticker tape timer.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
For Use With (Equipment) Photogate and Bar Tape
Length (Metric) 3 m