Learn More
Advanced Biomatrix PhotoGel Methacrylated Gelatin 50% Degree of methacrylation 1 gram (2 x 500 mg)
Catalog No. 502255956
Encompass_Preferred
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness.
This item is not returnable. View return policy
Catalog No. 50-225-5956 Supplier Advanced Biomatrix Supplier No. VL3500000502
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness.