Learn More
Advanced Biomatrix PhotoGel 50% DOM Methacrylated Gelatin with Irgacure Kit
Catalog No. NC2366532
Encompass_Preferred
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin and a photoinitiator. The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. The photoinitiator consists of Irgacure 2959 to be formulated in methanol (methanol not included) which allows UV crosslinking of the gelatin at 365 nm.
Catalog No. NC2366532 Supplier Advanced Biomatrix Supplier No. 53981KIT
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin and a photoinitiator. The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. The photoinitiator consists of Irgacure 2959 to be formulated in methanol (methanol not included) which allows UV crosslinking of the gelatin at 365 nm.