Learn More
Advanced Biomatrix PhotoGel 50% DOM Methacrylated Gelatin with LAP Kit
Catalog No. NC2366533
Encompass_Preferred
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin and a photoinitiator. The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. The photoinitiator consists of LAP which allows visible light crosslinking of the gelatin at 405 nm.
Catalog No. NC2366533 Supplier Advanced Biomatrix Supplier No. 53991KIT
PhotoGel is lyophilized, sterile, medium degree of methacrylation (~50%) methacrylated gelatin. PhotoGel provides 3D gelatin hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin and a photoinitiator. The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. The photoinitiator consists of LAP which allows visible light crosslinking of the gelatin at 405 nm.