Learn More
Advanced Biomatrix PhotoGel-LAP Methacrylated Gelatin + LAP Kit
Catalog No. 501960310
Encompass_Preferred
Advanced BioMatrix offers PhotoGel LAP Kit, a purified (sterility tested) methacrylated gelatin kit for photocrosslinkable hydrogels. PhotoGelprovides 3D tunable gels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin (1 gram) and a LAP Photoinitiator (405 nm). The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the -amines groups of the lysine residues as well as the a-amines groups on the N-termini. >75% of the total lysine residues of the gelatin molecule have been methacrylated. The photoinitiator consists of LAP to be formulated in 1X PBS or cell culture media which allows blue light photocrosslinking of the gelatin at 405 nm.
Catalog No. 50-196-0310 Supplier Advanced Biomatrix Supplier No. 52721KIT
Advanced BioMatrix offers PhotoGel LAP Kit, a purified (sterility tested) methacrylated gelatin kit for photocrosslinkable hydrogels. PhotoGelprovides 3D tunable gels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. The PhotoGel kit consists of purified methacrylated porcine gelatin (1 gram) and a LAP Photoinitiator (405 nm). The gelatin is type A 300 bloom. PhotoGel is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the -amines groups of the lysine residues as well as the a-amines groups on the N-termini. >75% of the total lysine residues of the gelatin molecule have been methacrylated. The photoinitiator consists of LAP to be formulated in 1X PBS or cell culture media which allows blue light photocrosslinking of the gelatin at 405 nm.