Learn More
Advanced Biomatrix PhotoGel-Ink methacrylated gelatin bioink for BIONOVA X DLP Printer 25 mL 95% degree of methacrylation
Catalog No. 502255966
Encompass_Preferred
This PhotoGel-INK is a sterile, ready-to-use 95% degree of methacrylation gelatin (10% solution), with 0.25% LAP, designed to work flawlessly with the BIONOVA X DLP Bioprinter for innovative 3D cell culture.
This item is not returnable. View return policy
Catalog No. 50-225-5966 Supplier Advanced Biomatrix Supplier No. D16110025360
This PhotoGel-INK is a sterile, ready-to-use 95% degree of methacrylation gelatin (10% solution), with 0.25% LAP, designed to work flawlessly with the BIONOVA X DLP Bioprinter for innovative 3D cell culture.