Learn More
Advanced Biomatrix PhotoGel-IRG Methacrylated Gelatin + Irgacure Kit
Catalog No. NC1407571
Encompass_Preferred
Advanced BioMatrix offers PhotoGel Irgacure Kit, a purified methacrylated gelatin kit for UV light (365 nm) photocrosslinkable hydrogels. PhotoGel provides 3D tunable gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness. PhotoGel is produced from methacrylated gelatin (1 gram) where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. >75% of the total lysine residues of the gelatin molecule have been methacrylated. The photoinitiator consists of Irgacure to be formulated in methanol which allows visible light photocrosslinking of the gelatin at 365 nm.
Catalog No. NC1407571 Supplier Advanced Biomatrix Supplier No. 52151KIT
Advanced BioMatrix offers PhotoGel Irgacure Kit, a purified methacrylated gelatin kit for UV light (365 nm) photocrosslinkable hydrogels. PhotoGel provides 3D tunable gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness. PhotoGel is produced from methacrylated gelatin (1 gram) where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. >75% of the total lysine residues of the gelatin molecule have been methacrylated. The photoinitiator consists of Irgacure to be formulated in methanol which allows visible light photocrosslinking of the gelatin at 365 nm.