Learn More
Advanced Biomatrix PhotoHA-RUT Methacrylated Hyaluronic Acid + Ruthenium Kit
Catalog No. 501960313
Encompass_Preferred
Advanced BioMatrix offers PhotoHA, a methacrylated hyaluronic acid (HAMA) for photocrosslinkable hydrogels. These hydrogels provide native-like 3D HA gels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. The kit comes with 100 mg of lyophilized methacrylated hyaluronic acid and ruthenium and sodium persulfate photoinitiators (400-450 nm visible light photocrosslinking). Hyaluronic acid is the most abundant glycosaminoglycan in the body being an important component of several tissues throughout the body. While it is abundant in extracellular matrices, hyaluronic acid also contributes to tissue hydrodynamics, movement and proliferation of cells, and participates in a number of cell surface receptor interactions.
Catalog No. 50-196-0313 Supplier Advanced Biomatrix Supplier No. 52751KIT
Advanced BioMatrix offers PhotoHA, a methacrylated hyaluronic acid (HAMA) for photocrosslinkable hydrogels. These hydrogels provide native-like 3D HA gels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. The kit comes with 100 mg of lyophilized methacrylated hyaluronic acid and ruthenium and sodium persulfate photoinitiators (400-450 nm visible light photocrosslinking). Hyaluronic acid is the most abundant glycosaminoglycan in the body being an important component of several tissues throughout the body. While it is abundant in extracellular matrices, hyaluronic acid also contributes to tissue hydrodynamics, movement and proliferation of cells, and participates in a number of cell surface receptor interactions.