Learn More
Advanced Biomatrix PhotoHA-Ink Soft methacrylated hyaluronic acid bioink for BIONOVA X DLP Printer 5 mL
Catalog No. 502255964
Encompass_Preferred
PhotoHA-INK (soft) is 5 mL of a 4% solution of methacrylated hyaluronic acid (GM-HA) with 0.5% LAP. PhotoHA (soft) is a GM-HA which provides softer hydrogels than the (stiff) HAMA. This ink is pre-formulated and ready for cell printing in the BIONOVA X.
This item is not returnable. View return policy
Catalog No. 50-225-5964 Supplier Advanced Biomatrix Supplier No. D16110025376
PhotoHA-INK (soft) is 5 mL of a 4% solution of methacrylated hyaluronic acid (GM-HA) with 0.5% LAP. PhotoHA (soft) is a GM-HA which provides softer hydrogels than the (stiff) HAMA. This ink is pre-formulated and ready for cell printing in the BIONOVA X.