Learn More
Advanced Biomatrix PhotoHA-Ink Stiff methacrylated hyaluronic acid bioink for BIONOVA X DLP Printer 5 mL
Catalog No. 502255963
Encompass_Preferred
PhotoHA-INK (stiff) is 5 mL of a 4% solution of methacrylated hyaluronic acid (HAMA) with 0.5% LAP, and forms stronger hydrogels than the PhotoHA-INK (soft), which is a GM-HA. This Ink is pre-formulated and ready for printing in the BIONOVA X.
This item is not returnable. View return policy
Catalog No. 50-225-5963 Supplier Advanced Biomatrix Supplier No. D16110025362
PhotoHA-INK (stiff) is 5 mL of a 4% solution of methacrylated hyaluronic acid (HAMA) with 0.5% LAP, and forms stronger hydrogels than the PhotoHA-INK (soft), which is a GM-HA. This Ink is pre-formulated and ready for printing in the BIONOVA X.