Learn More
Advanced Biomatrix PhotoHA-Soft Methacrylated hyaluronic acid softer hydrogels 100 mg
Catalog No. 502255969
Encompass_Preferred
PhotoHA is 100 mg of lyophilized methacrylated hyaluronic acid (GM-HA). PhotoHA provides 3D hyaluronic acid hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. GM-HA forms softer hydrogels than HAMA.
This item is not returnable. View return policy
Catalog No. 50-225-5969 Supplier Advanced Biomatrix Supplier No. 5388100MG
PhotoHA is 100 mg of lyophilized methacrylated hyaluronic acid (GM-HA). PhotoHA provides 3D hyaluronic acid hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. GM-HA forms softer hydrogels than HAMA.