Learn More
LOGOS BIOSYSTEMS INC PhotonSlideTM 50 Slides
SDP
Catalog No. NC1591330
Encompass
The PhotonSlide 50 Slides are microscope slides with a hydrophobic surface that allows for uniform spreading of small liquid samples. They are made of soda lime glass and measure 25x75x1mm. The slides have frosted writing areas for labeling and identification. They come in a pack of 50 slides and are suitable for use with most microscope slide staining procedures. The hydrophobic surface allows for even spreading of samples as small as 5 microliters.
Catalog No. NC1591330 Supplier LOGOS BIOSYSTEMS INC Supplier No. L12005
The PhotonSlide 50 Slides are microscope slides with a hydrophobic surface that allows for uniform spreading of small liquid samples. They are made of soda lime glass and measure 25x75x1mm. The slides have frosted writing areas for labeling and identification. They come in a pack of 50 slides and are suitable for use with most microscope slide staining procedures. The hydrophobic surface allows for even spreading of samples as small as 5 microliters.