Description
This Photooxidation UV Lamp degrades organic compounds in ultrapure water, reducing the TOC (total organic carbon) to less than 5 parts per billion.
- For use with Milli-Q™ EQ 7000 water purification systems
- 185 nm mercury UV lamp
- Photooxidative effect
- Easy replacement
Specifications
Specifications
| Product Type | UV Lamp |
| For Use With (Equipment) | Milli-Q™ EQ 7000 Water Purification Systems |
| Wavelength | 185 nm |