Learn More
Vector Laboratories PHOTOPROBE (Long Arm) Biotin Kit for Nucleic Acid Labeling
SDP
Catalog No. NC9105886
Encompass
PHOTOPROBE (Long Arm) Biotin is used to incorporate biotin at multiple sites along a nucleic acid without destroying or copying the original nucleic acid. It can be used to label DNA, RNA or PNA. The labeling reagent has a long linker arm to increase the accessibility of the biotin. Each kit contains PHOTOPROBE Biotin Reagent, Tris Buffer, sec-Butanol Precipitant, and Biotinylated DNA Standard, and can be used to label up to 250 µg of target or for up to 50 labeling reactions.
Catalog No. NC9105886 Supplier Vector Laboratories Supplier No. SP1020.5
PHOTOPROBE (Long Arm) Biotin is used to incorporate biotin at multiple sites along a nucleic acid without destroying or copying the original nucleic acid. It can be used to label DNA, RNA or PNA. The labeling reagent has a long linker arm to increase the accessibility of the biotin. Each kit contains PHOTOPROBE Biotin Reagent, Tris Buffer, sec-Butanol Precipitant, and Biotinylated DNA Standard, and can be used to label up to 250 µg of target or for up to 50 labeling reactions.