Learn More
HEPATOCHEM PhotoRedOx Box UV-Vis Photoreactor
Catalog No. NC3714381
Encompass
This photoreactor is designed to perform photochemical reactions using UV and Visible light from 254 nm to 808 nm. It is compatible with holders for 8X 4 ml vials, 8X 8ml vials and 4X 20 ml vials. The reaction temperature is monitored using a IR probe and regulated with internal fans. This photoreactor comprises a stirring module and user interface to setup the experiments.
Catalog No. NC3714381 Supplier HEPATOCHEM Supplier No. HCK102201001
Add to Cart
Add to Cart
Request a Quote
This photoreactor is designed to perform photochemical reactions using UV and Visible light from 254 nm to 808 nm. It is compatible with holders for 8X 4 ml vials, 8X 8ml vials and 4X 20 ml vials. The reaction temperature is monitored using a IR probe and regulated with internal fans. This photoreactor comprises a stirring module and user interface to setup the experiments.