Learn More
HEPATOCHEM Flow reactor 2 ml with 1/16 tubing for PhotoRedOx Box, Duo, and TC
Catalog No. NC2184648
Encompass_Preferred
2 ml flow reactor with 1/16 PFA tubing for PhotoRedOx Box™ Single HCK1006-01-016, PhotoRedOx Box™ Duo HCK1006-01-023, and PhotoRedOx Box™ TC HCK1006-01-025
This item is not returnable. View return policy
Catalog No. NC2184648 Supplier HEPATOCHEM Supplier No. HCK100601022
2 ml flow reactor with 1/16 PFA tubing for PhotoRedOx Box™ Single HCK1006-01-016, PhotoRedOx Box™ Duo HCK1006-01-023, and PhotoRedOx Box™ TC HCK1006-01-025