Learn More
HEPATOCHEM EvoluChem PhotoRedOx Box™ photoreactor TC
Catalog No. NC1973633
Encompass_Preferred
This temperature controlled photoreactor (US Patent #10,906,022) enables photo-catalytic reactions at controlled temperatures between 0°C to 80°C. A flexible design allows interchangeable LEDs from 365 nm to 808 nm and a wide variety of vials. The design is based on our original PhotoRedOx Box™ which features a unique reaction chamber geometry that directs light throughout the chamber, allowing the performance of multiple reaction conditions simultaneously (between 2 and 32 depending on the reaction vial sizes). However, with the aluminum-based, waterproof PhotoRedOx TC™, it is possible to heat and cool the reaction medium using a thermostatic fluid (such as water or ethylene glycol) that recirculates through a standard chiller/heater unit.
Catalog No. NC1973633 Supplier HEPATOCHEM Supplier No. HCK100601025
This temperature controlled photoreactor (US Patent #10,906,022) enables photo-catalytic reactions at controlled temperatures between 0°C to 80°C. A flexible design allows interchangeable LEDs from 365 nm to 808 nm and a wide variety of vials. The design is based on our original PhotoRedOx Box™ which features a unique reaction chamber geometry that directs light throughout the chamber, allowing the performance of multiple reaction conditions simultaneously (between 2 and 32 depending on the reaction vial sizes). However, with the aluminum-based, waterproof PhotoRedOx TC™, it is possible to heat and cool the reaction medium using a thermostatic fluid (such as water or ethylene glycol) that recirculates through a standard chiller/heater unit.