Learn More
Caron Products And Services Photostability Chamber, 11 cu. ft., Option 2, Temp
Supplier:  Caron Products And Services 7540111
Encompass_Preferred
Photostability Chamber, 11 cu. ft. Meets ICH Q1B, Option 2, requirements with Cool White and Near UV lamps. Temperature controlled from 10C to 35C. Electrical: 115V, 60 Hz, 15A.
Catalog No. NC1830017
Add to Cart
Request a Quote