Learn More
Caron Products And Services Photostability Chamber, 11 cu. ft., Option 2, Temp & RH
Catalog No. NC2386461
Encompass_Preferred
Photostability Chamber, 11 cu. ft. Meets ICH Q1B, Option 2, requirements with Cool White and Near UV lamps. Temperature controlled from 10C to 35C and humidity controlled from 40% to 70% RH. Electrical: 208/230V, 60 Hz, 15A.
Catalog No. NC2386461 Supplier Caron Products And Services Supplier No. 7545112
Add to Cart
Add to Cart
Request a Quote
Photostability Chamber, 11 cu. ft. Meets ICH Q1B, Option 2, requirements with Cool White and Near UV lamps. Temperature controlled from 10C to 35C and humidity controlled from 40% to 70% RH. Electrical: 208/230V, 60 Hz, 15A.